- Determination of deslanoside in antemortem and postmortem specimens. Unusual case report.
Determination of deslanoside in antemortem and postmortem specimens. Unusual case report.
Forensic science international (1990-04-01)
M Klys, A Gross
PMID2193860
ABSTRACT
One case of the erroneous administration of deslanoside and high level of drug in antemortem plasma and postmortem specimens has been reported owing to the unusual surrounding circumstances. Deslanoside in antemortem plasma was determined by FPIA and the analysis was done by HPLC in the postmortem tissue samples. The analytical results and methods used in the examinations are discussed in the following paper.
Product Description
Deslanoside, European Pharmacopoeia (EP) Reference Standard