Merck
  • [Interferon inductor cupping of postvaccinal complications after variolation].
[Interferon inductor cupping of postvaccinal complications after variolation].
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic] (2010-06-30)
S Ia Loginova, S V Borisevich, V A Maksimov, V P Bondarev, V V Perekrest, A M Shuster
ABSTRACT
Efficacy of arbidol and ridostin in cupping postvaccinal complications due to variolation was studied by the clinico-virological, hematological and biochemical indices and it was shown that arbidol was efficient in cupping development of dermal complications, lowered the severity of the postvaccinal reaction and stimulated the cellular and humoral immune response. Ridostin, a high molecular interferon inductor, was highly efficient in cupping all the forms of the postvaccinal complications, including the neurological and cutaneous ones.
Sigma-Aldrich
Arbidol hydrochloride, ≥98% (HPLC)