Merck
  • Isolation and identification of antifungal and antialgal alkaloids from Haplophyllum sieversii.
Isolation and identification of antifungal and antialgal alkaloids from Haplophyllum sieversii.
Journal of agricultural and food chemistry (2005-09-30)
C L Cantrell, K K Schrader, L K Mamonov, G T Sitpaeva, T S Kustova, C Dunbar, D E Wedge
ABSTRACT
Bioassay-guided fractionation of the hexane/ethyl acetate/water (H/EtOAc/H2O) crude extract of the aerial parts of Haplophyllum sieversii was performed because of preliminary screening data that indicated the presence of growth inhibitory components against Colletotrichum fragariae, Colletotrichum gloeosporioides, and Colletotrichum acutatum. Fractionation was directed using bioautographical methods resulting in the isolation of the bioactive alkaloids flindersine, anhydroevoxine, haplamine, and a lignan eudesmin. These four compounds were evaluated for activity against C. fragariae, C. gloeosporioides, C. acutatum, Botrytis cinerea, Fusarium oxysporum, and Phomopsis obscurans in a dose-response growth-inhibitory bioassay at 50.0, 100.0, and 150.0 microM. Of the four compounds tested, flindersine demonstrated the highest level of antifungal activity. Additionally, flindersine, eudesmin, and haplamine were screened against the freshwater phytoplanktons Oscillatoria perornata, Oscillatoria agardhii, Selenastrum capricornutum, and Pseudanabaena sp. (strain LW397). Haplamine demonstrated selective inhibition against the odor-producing cyanobacterium O. perornata compared to the activity against the green alga S. capricornutum, with lowest observed effect concentration values of 1.0 and 10.0 microM, respectively.
Sigma-Aldrich
Eudesmine, ≥95% (LC/MS-ELSD)