Merck
Regulation of ceramide production and apoptosis.
Annual review of physiology (1998-04-29)
R N Kolesnick, M Krönke
ABSTRACT
Ceramide is a sphingosine-based lipid signaling molecule that regulates cellular differentiation, proliferation, and apoptosis. The emerging picture suggests that coupling of ceramide to specific signaling cascades is both stimulus and cell-type specific. Ceramide action is determined within the context of other stimuli and by the subcellular topology of its production. Here, we discuss the pathways of ceramide generation and the interaction of ceramide with caspases and other apoptotic signaling cascades.
Sigma-Aldrich
Anti-SMPD2 antibody produced in rabbit, IgG fraction of antiserum