- Photochemical polymerization of thiophene derivatives in aqueous solution.
Photochemical polymerization of thiophene derivatives in aqueous solution.
Chemical communications (Cambridge, England) (2004-10-07)
Sergey A Piletsky, Elena V Piletska, Kal Karim, Frank Davis, Seamus P J Higson, Anthony P F Turner
PMID15467882
ABSTRACT
A novel method of photochemical polymerization of thiophene derivatives in aqueous solution catalyzed by potassium dichromate and initiated by illumination is described.
MATERIALS