Merck
  • cTAGE5 mediates collagen secretion through interaction with TANGO1 at endoplasmic reticulum exit sites.
cTAGE5 mediates collagen secretion through interaction with TANGO1 at endoplasmic reticulum exit sites.
Molecular biology of the cell (2011-04-29)
Kota Saito, Koh Yamashiro, Yuki Ichikawa, Patrik Erlmann, Kenji Kontani, Vivek Malhotra, Toshiaki Katada
ABSTRACT
Cutaneous T-cell lymphoma-associated antigen 5 (cTAGE5), an originally identified tumor antigen, is overexpressed in various cancer cell lines. The cDNA encodes an integral membrane protein containing two coiled-coil motifs and a proline-rich domain. We show that cTAGE5 specifically localizes to the endoplasmic reticulum (ER) exit sites. In addition, cTAGE5 forms a complex with TANGO1 (MIA3), a previously characterized cargo receptor for collagen VII, by the interaction of their coiled-coil motifs. Of interest, cTAGE5, as well as TANGO1, is capable of interacting with the inner-layer coatomer of COPII Sec23/24 complex through their C-terminal proline-rich domains and required for collagen VII secretion. We propose that cTAGE5 acts as a coreceptor of TANGO1 for collagen VII export from the ER.