Merck
  • Seasonal and sex-related variations in serum steroid hormone levels in wild and farmed brown trout Salmo trutta L. in the north-west of Spain.
Seasonal and sex-related variations in serum steroid hormone levels in wild and farmed brown trout Salmo trutta L. in the north-west of Spain.
Journal of water and health (2013-12-18)
Juan M Fregeneda-Grandes, Salvador Hernández-Navarro, Ignacio A Fernandez-Coppel, Adriana Correa-Guimaraes, Norlan Ruíz-Potosme, Luis M Navas-Gracia, J Miguel Aller-Gancedo, Francisco J Martín-Gil, Jesús Martín-Gil
ABSTRACT
Serum steroid profiles were investigated in order to evaluate the potential use of circulating sex steroid levels as a tool for sex identification in brown trout. Changes in the serum concentrations of testosterone (T), progesterone (P), 17-β-estradiol (E2), and cortisol (F) in wild and farmed mature female and male brown trout, Salmo trutta L., were measured in each season (January, May, July, and October) in six rivers and four hatcheries located in the north-west of Spain. Serum cortisol levels in farmed brown trout were significantly higher and showed a seasonal pattern opposite to that found in wild trout. Because levels of the hormones under study can be affected by disruptive factors such as exposure to phytoestrogens (which alters the hypothalamic-pituitary-gonadal axis) and infection with Saprolegnia parasitica (which alters the hypothalamic-pituitary-adrenal axis), both factors are taken into account.
Product Description
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Progesterone, meets USP testing specifications
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard