Merck
  • Neuroestradiol in the hypothalamus contributes to the regulation of gonadotropin releasing hormone release.
Neuroestradiol in the hypothalamus contributes to the regulation of gonadotropin releasing hormone release.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-12-07)
Brian P Kenealy, Amita Kapoor, Kathryn A Guerriero, Kim L Keen, James P Garcia, Joseph R Kurian, Toni E Ziegler, Ei Terasawa
ABSTRACT
Release of gonadotropin releasing hormone (GnRH) from the medial basal hypothalamus (MBH)/median eminence region (S-ME) is essential for normal reproductive function. GnRH release is profoundly regulated by the negative and positive feedback effects of ovarian estradiol (E2). Here we report that neuroestradiol, released in the S-ME, also directly influences GnRH release in ovariectomized female monkeys, in which the ovarian source of E2 is removed. We found that (1) brief infusion of E2 benzoate (EB) to the S-ME rapidly stimulated release of GnRH and E2 in the S-ME of ovariectomized monkeys, (2) electrical stimulation of the MBH resulted in GnRH release as well as E2 release, and (3) direct infusion of an aromatase inhibitor to the S-ME suppressed spontaneous GnRH release as well as the EB-induced release of GnRH and E2. These findings reveal the importance of neuroestradiol as a neurotransmitter in regulation of GnRH release. How circulating ovarian E2 interacts with hypothalamic neuroestrogens in the control of GnRH release remains to be investigated.
Sigma-Aldrich
β-Estradiol 3-benzoate, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Estradiol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Letrozole, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Letrozole, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
β-Estradiol, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Estradiol, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Estradiol hemihydrate, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®