Merck
  • Modular isoquinoline synthesis using catalytic enolate arylation and in situ functionalization.
Modular isoquinoline synthesis using catalytic enolate arylation and in situ functionalization.
Organic letters (2013-11-21)
Ben S Pilgrim, Alice E Gatland, Charlie T McTernan, Panayiotis A Procopiou, Timothy J Donohoe
ABSTRACT
A methyl ketone, an aryl bromide, an electrophile, and ammonium chloride were combined in a four-component, three-step, and one-pot coupling procedure to furnish substituted isoquinolines in overall yields of up to 80%. This protocol utilizes the palladium catalyzed α-arylation reaction of an enolate, followed by in situ trapping with an electrophile, and aromatization with ammonium chloride. tert-Butyl cyanoacetate participated in a similar protocol; after functionalization and decarboxylation, 3-amino-4-alkyl isoquinolines were prepared in high yield.
Product Description
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
tert-Butyl cyanoacetate, ≥97.0% (GC)
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Isoquinoline, 97%
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis