- High temperature oxidation of ZrO2/Al2O3 thin films deposited on steel.
High temperature oxidation of ZrO2/Al2O3 thin films deposited on steel.
Journal of nanoscience and nanotechnology (2013-11-20)
Jae Chun Lee, Sun Kyu Kim, Trinh Van Trung, Dong Bok Lee
PMID24245292
ABSTRACT
Thin ZrO2/Al2O3 films that consisted of alternating monoclinic ZrO2 nanolayers and amorphous Al2O3 nanolayers were deposited on a tool steel substrate using Zr and Al cathodes in a cathodic arc plasma deposition system, and then oxidized at 600-900 degrees C in air for up to 50 h. The ZrO2/Al2O3 films effectively suppressed the oxidation of the substrate up to 800 degrees C by acting as a barrier layer against the outward diffusion of the substrate elements and inward diffusion of oxygen. However, rapid oxidation occurred at 900 degrees C due mainly to the increased diffusion and subsequent oxidation of steel as well as the crystallization of amorphous Al2O3.
Product Description
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O