Merck
  • Embryonic exposure of medaka (Oryzias latipes) to propylparaben: effects on early development and post-hatching growth.
Embryonic exposure of medaka (Oryzias latipes) to propylparaben: effects on early development and post-hatching growth.
Environmental pollution (Barking, Essex : 1987) (2013-10-08)
Miguel González-Doncel, José Enrique García-Mauriño, Laura San Segundo, Eulalia M Beltrán, Salvador Sastre, Carlos Fernández Torija
ABSTRACT
Here we proposed a battery of non-invasive biomarkers and a histological survey to examine physiological/anatomical features in embryos, eleutheroembryos (13 days post-fertilization, dpf), and larvae (28-42 dpf) of medaka to investigate the effects of embryonic exposure to propylparaben (PrP). Concentrations <1000 μg PrP/L didn't exert early or late toxic effects. However, survivorship was affected at 4000 μg/L in eleutheroembryos and at ≥1000 μg/L in larvae. Histological alterations were found in 37.5% of eleutheroembryos exposed to 4000 μg PrP/L. Morphometric analysis of the gallbladder revealed significant dilation at ≥400 μg/L throughout embryo development. Ethoxyresorufin-O-deethylase (EROD), as indicator of cytochrome P4501A activity, didn't reveal induction/inhibition although its combination with a P4501A agonist (i.e. β-naphthoflavone) resulted in a synergic EROD response. Results suggest a low toxicity of PrP for fish and support the use of fish embryos and eleutheroembryos as alternatives of in vivo biomarkers indicative of exposure/toxicity.
Product Description
Supelco
Propylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Propyl parahydroxybenzoate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, BioXtra
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl 4-hydroxybenzoate, ≥99%
Pricing and availability is not currently available.