Merck
  • Potentiation of antibiofilm activity of amphotericin B by superoxide dismutase inhibition.
Potentiation of antibiofilm activity of amphotericin B by superoxide dismutase inhibition.
Oxidative medicine and cellular longevity (2013-10-01)
Katrijn De Brucker, Anna Bink, Els Meert, Bruno P A Cammue, Karin Thevissen
ABSTRACT
This study demonstrates a role for superoxide dismutases (Sods) in governing tolerance of Candida albicans biofilms to amphotericin B (AmB). Coincubation of C. albicans biofilms with AmB and the Sod inhibitors N,N'-diethyldithiocarbamate (DDC) or ammonium tetrathiomolybdate (ATM) resulted in reduced viable biofilm cells and increased intracellular reactive oxygen species levels as compared to incubation of biofilm cells with AmB, DDC, or ATM alone. Hence, Sod inhibitors can be used to potentiate the activity of AmB against C. albicans biofilms.
Product Description
Supelco
Sodium diethyldithiocarbamate trihydrate, ACS reagent, for the determination and separation of heavy metals, ≥97%
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate, ACS reagent
Sigma-Aldrich
Ammonium diethyldithiocarbamate
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard