Merck
  • Evaluation of diffusive gradients in thin films technique (DGT) for measuring Al, Cd, Co, Cu, Mn, Ni, and Zn in Amazonian rivers.
Evaluation of diffusive gradients in thin films technique (DGT) for measuring Al, Cd, Co, Cu, Mn, Ni, and Zn in Amazonian rivers.
Environmental monitoring and assessment (2013-09-21)
Lauren Nozomi Marques Yabuki, Camila Destro Colaço, Amauri Antonio Menegário, Roberto Naves Domingos, Chang Hung Kiang, Domitila Pascoaloto
ABSTRACT
Studies concerning the lability and bioavailability of trace metals have played a prominent role in the search for contamination of water resources. This work describes the first application yet of the diffusive gradients in thin films technique (DGT) to the determination of the fraction of free plus labile metals in waters from the Amazon Basin. Due to the complexity of the use of DGT for samples with low ionic strength and high organic matter content (characteristic of Amazonian rivers), a new analytical procedure was developed. The method is based on the determinations of apparent diffusion coefficients (Dap) in the laboratory, by performing deployments in samples collected in the corresponding sites of study. The Dap thereby determined is then used for in situ measurements. The suitability of the proposed approach for determination of labile Al, Cd, Co, Cu, Mn, Ni, and Zn in the Amazon River and Rio Negro (English: Black River) was evaluated. Except for Co, Mn (in a deployment at Rio Negro), Ni and Zn (in a deployment at Amazon River), labile in situ measurements were lower or similar to dissolved concentrations, indicating suitability of the proposed approach.
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Nickel, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Sigma-Aldrich
Nickel, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Nickel, rod, diam. 6.35 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%