Merck
  • Indium chloride-induced micronuclei via reactive oxygen species in Chinese hamster lung fibroblast V79 cells.
Indium chloride-induced micronuclei via reactive oxygen species in Chinese hamster lung fibroblast V79 cells.
Environmental toxicology (2013-09-12)
Ruey-Hseng Lin, Ming-Ling Yang, Yi-Ching Li, Hui-Min Chang, Yu-Hsiang Kuan
ABSTRACT
We study the cytotoxicity of indium chloride (InCl₃) in Chinese hamster lung fibroblasts, the V79 cells, using MTT assay. The results showed that InCl₃ did not induce significant cytotoxicity at various concentrations tested. In addition, the frequency of micronuclei (MN) was assayed to evaluate the genotoxic effects of InCl₃ in V79 cells. InCl₃ at concentrations ranged 0.1-1 μM significantly increased MN frequency in a concentration-dependent manner. Both catalase and superoxide dismutase at concentrations of 75 and 150 μg/mL significantly inhibited InCl₃-induced MN. Similarly, Germanium oxide (GeO₂) and dimercaprol expressed antigenotoxic effects. From these findings, it is concluded that InCl₃ is a potent genotoxic chemical, which may be mediated partly by inducing oxidative stress. The significance of this study shows that the workers in the semiconductor factories should be cautious in exposing to the hazardous genotoxic InCl₃.
Sigma-Aldrich
Germanium(IV) oxide, (crystalline powder), 99.998% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Germanium(IV) oxide, powder, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 98%
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium(III) chloride, anhydrous, powder, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Germanium, chips, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Germanium, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Indium(III) chloride, 99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)