Merck
  • A novel drug delivery system of intraperitoneal chemotherapy for peritoneal carcinomatosis using gelatin microspheres incorporating cisplatin.
A novel drug delivery system of intraperitoneal chemotherapy for peritoneal carcinomatosis using gelatin microspheres incorporating cisplatin.
Surgery (2013-09-07)
Shutaro Gunji, Kazukata Obama, Makoto Matsui, Yasuhiko Tabata, Yoshiharu Sakai
ABSTRACT
Peritoneal carcinomatosis is a poor prognostic factor for patients with gastrointestinal, gynecologic, and pancreatic cancer. Cisplatin (CDDP) is among the most effective anti-cancer agents, although its adverse effects remain unresolved. For the treatment of peritoneal carcinomatosis with high-dose CDDP, it is necessary to design a new delivery system of CDDP that can decrease systemic toxicity and achieve a better targeted, high-dose chemotherapy. Microspheres were prepared from gelatin of a nontoxic, biodegradable material for the sustained release of CDDP. The gelatin microspheres incorporating CDDP (GM-CDDP) were injected intraperitoneally into a mouse model of peritoneal carcinomatosis; their therapeutic efficacy and adverse effects were evaluated in comparison with intraperitoneal administration of free CDDP. GM-CDDP released CDDP in the peritoneal cavity as a result of gelatin biodegradation. Mice treated with microspheres in the peritoneal cavity lived longer than mice treated with free CDDP (74 ± 23 vs 40 ± 23 days; P < .05). The mice treated with GM-CDDP also lost no weight, whereas the free CDDP group lost approximately 20% body weight (106 ± 5% vs 80 ± 7%; P < .001; body weight on day 1 = 100%). GM-CDDP significantly decreased the nephrotoxicity and hematotoxicity of CDDP. GM decreased the adverse effects of CDDP and allowed high-dose intraperitoneal chemotherapy with the control of CDDP. This technique of gradual local release may allow us to provide a high-dose, targeted, intraperitoneal chemotherapy with CDDP, resulting in enhanced anti-cancer effects. These gelatin microspheres may be useful as a drug carrier for the treatment of peritoneal carcinomatosis.
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from cold water fish skin, solid