Merck
  • Allomorph distribution and granule structure of lotus rhizome C-type starch during gelatinization.
Allomorph distribution and granule structure of lotus rhizome C-type starch during gelatinization.
Food chemistry (2013-09-05)
Canhui Cai, Jinwen Cai, Jianmin Man, Yang Yang, Zhifeng Wang, Cunxu Wei
ABSTRACT
The allomorph distribution and granule structure of C-type starch from lotus rhizomes were investigated using a combination of techniques during gelatinization. The disruption of crystallinity during gelatinization began from the end distant from the eccentric hilum and then propagated into the center of granule. The periphery of hilum end was finally gelatinized, accompanied by high swelling. The crystallinity changed from C-type to A-type via CA-type during gelatinization, and finally became amorphous structure. The amylose content, crystal degree, helix content, ratio of 1045/1022cm(-1), and peak intensity of crystalline lamellae of gelatinizing starch significantly decreased after 70°C. The amorphous content and ratio of 1022/995cm(-1) increased after 70°C. This study elucidated that B-type allomorph was mainly arranged in the distal region of eccentric hilum, A-type allomorph was mainly located in the periphery of hilum end, and the center of granule was a mixed distribution of A- and B-type allomorphs.
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture