Merck
An unusual cause of persistent postpartum vaginal discharge.
International journal of STD & AIDS (2013-08-24)
Natasha Parrott, Maria Asensio, Nadi K Gupta
ABSTRACT
We describe the case of a 27-year-old woman with a three-month history of persistent green vaginal discharge following vaginal delivery of her first child. Group C β-haemolytic streptococcus was ultimately isolated, which appears to be an uncommon cause of this clinical presentation.
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard