Merck
  • Targeting the sphingosine-1-phosphate axis in cancer, inflammation and beyond.
Targeting the sphingosine-1-phosphate axis in cancer, inflammation and beyond.
Nature reviews. Drug discovery (2013-08-21)
Gregory T Kunkel, Michael Maceyka, Sheldon Milstien, Sarah Spiegel
ABSTRACT
The bioactive lipid sphingosine-1-phosphate (S1P) is involved in multiple cellular signalling systems and has a pivotal role in the control of immune cell trafficking. As such, S1P has been implicated in disorders such as cancer and inflammatory diseases. This Review discusses the ways in which S1P might be therapeutically targeted - for example, via the development of chemical inhibitors that target the generation, transport and degradation of S1P and via the development of specific S1P receptor agonists. We also highlight recent conflicting results observed in preclinical studies targeting S1P and discuss ongoing clinical trials in this field.
Sigma-Aldrich
Sphingosine 1-phosphate, ≥98.0% (TLC)
Sigma-Aldrich
Sphingosine 1-phosphate, ≥95%, powder