Merck
  • Synthesis of 2-nitroglycals from glycals using the tetrabutylammonium nitrate-trifluoroacetic anhydride-triethylamine reagent system and base-catalyzed Ferrier rearrangement of acetylated 2-nitroglycals.
Synthesis of 2-nitroglycals from glycals using the tetrabutylammonium nitrate-trifluoroacetic anhydride-triethylamine reagent system and base-catalyzed Ferrier rearrangement of acetylated 2-nitroglycals.
The Journal of organic chemistry (2013-08-13)
Suresh Dharuman, Preeti Gupta, Pavan K Kancharla, Yashwant D Vankar
ABSTRACT
A reagent system comprising tetrabutylammonium nitrate-trifluoroacetic anhydride-triethylamine has been developed for the synthesis of 2-nitroglycals from various protected glycals. The base-catalyzed Ferrier rearrangement on tri-O-acetylated 2-nitroglycals has been reported for the first time. Reactivity of these nitroacetates and associated selectivity has been examined, and some of the products have been converted into 2,3-diamino-2,3-dideoxyglycosides and methyl N-acetyl-D-lividosaminide.
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O