Merck
  • Removal of 8-quinolinecarboxylic acid pesticide from aqueous solution by adsorption on activated montmorillonites.
Removal of 8-quinolinecarboxylic acid pesticide from aqueous solution by adsorption on activated montmorillonites.
Environmental monitoring and assessment (2013-08-09)
M Mekhloufi, A Zehhaf, A Benyoucef, C Quijada, E Morallon
ABSTRACT
Sodium montmorillonite (Na-M), acidic montmorillonite (H-M), and organo-acidic montmorillonite (Org-H-M) were applied to remove the herbicide 8-quinolinecarboxylic acid (8-QCA). The montmorillonites containing adsorbed 8-QCA were investigated by transmission electron microscopy, FT-IR spectroscopy, X-ray diffraction analysis, X-ray fluorescence thermogravimetric analysis, and physical adsorption of gases. Experiments showed that the amount of adsorbed 8-QCA increased at lower pH, reaching a maximum at pH 2. The adsorption kinetics was found to follow the pseudo-second-order kinetic model. The Langmuir model provided the best correlation of experimental data for adsorption equilibria. The adsorption of 8-QCA decreased in the order Org-H-M > H-M > Na-M. Isotherms were also used to obtain the thermodynamic parameters. The negative values of ΔG indicated the spontaneous nature of the adsorption process.
Sigma-Aldrich
Nanoclay, hydrophilic bentonite
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Bentonite