Merck
  • The effect of magnesium sulphate infusion on the incidence and severity of emergence agitation in children undergoing adenotonsillectomy using sevoflurane anaesthesia.
The effect of magnesium sulphate infusion on the incidence and severity of emergence agitation in children undergoing adenotonsillectomy using sevoflurane anaesthesia.
Anaesthesia (2013-08-06)
M Abdulatif, A Ahmed, A Mukhtar, S Badawy
ABSTRACT
This randomised, controlled, double-blind study investigated the effects of intra-operative magnesium sulphate administration on the incidence of emergence agitation in children undergoing adenotonsillectomy using sevoflurane anaesthesia. Seventy children were randomly allocated to receive a 30 mg.kg(-1) bolus of intravenous magnesium sulphate after induction of anaesthesia followed by a continuous infusion of 10 mg.kg(-1).h(-1) or an equal volume of saline 0.9%. All children received titrated sevoflurane anaesthesia adjusted to maintain haemodynamic stability. The Pediatric Anesthesia Emergence Delirium scale and the Children's Hospital of Eastern Ontario Score were used for the assessment of postoperative emergence agitation and pain, respectively. Emergence agitation was more common in the control group than in the magnesium group (23 (72%) and 12 (36%), respectively (p = 0.004)), with a relative risk of 0.51 (95% CI 0.31-0.84), an absolute risk reduction of 0.35 (95% CI 0.10-0.54), and number needed to treat of 3 (95% CI 2-9). Postoperative pain scores were comparable in the two groups. Magnesium sulphate reduces the incidence and severity of emergence agitation in children undergoing adenotonsillectomy using sevoflurane anaesthesia and is not associated with increased postoperative side-effects or delayed recovery.
Product Description
Sevoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%