Merck
  • Novel systemically active galanin receptor 2 ligands in depression-like behavior.
Novel systemically active galanin receptor 2 ligands in depression-like behavior.
Journal of neurochemistry (2013-04-23)
Indrek Saar, Jaanus Lahe, Kent Langel, Johan Runesson, Kristin Webling, Jaak Järv, Jussi Rytkönen, Ale Närvänen, Tamas Bartfai, Kaido Kurrikoff, Ülo Langel
ABSTRACT
Neuropeptide galanin and its three G-protein coupled receptors, galanin receptor type 1-galanin receptor type 3 (GalR1-GalR3), are involved in the regulation of numerous physiological and disease processes, and thus represent tremendous potential in neuroscience research and novel drug lead development. One of the areas where galanin is involved is depression. Previous studies have suggested that activation of GalR2 leads to attenuation of depression-like behavior. Unfortunately, lack of in vivo usable subtype specific ligands hinders testing the role of galanin in depression mechanisms. In this article, we utilize an approach of increasing in vivo usability of peptide-based ligands, acting upon CNS. Thus, we have synthesized a series of novel systemically active galanin analogs, with modest preferential binding toward GalR2. We have shown that specific chemical modifications to the galanin backbone increase brain levels upon i.v. injection of the peptides. Several of the new peptides, similar to a common clinically used antidepressant medication imipramine, exerted antidepressant-like effect in forced swim test, a mouse model of depression, at a surprisingly low dose range (< 0.5 mg/kg). We chose one of the peptides, J18, for more thorough study, and showed its efficacy also in another mouse depression model (tail suspension test), and demonstrated that its antidepressant-like effect upon i.v. administration can be blocked by i.c.v. galanin receptor antagonist M35. The effect of the J18 was also abolished in GalR2KO animals. All this suggests that systemically administered peptide analog J18 exerts its biological effect through activation of GalR2 in the brain. The novel galanin analogs represent potential drug leads and a novel pharmaceutical intervention for depression.
Product Description
Imipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imipramine hydrochloride, ≥99% (TLC)
Supelco
Imipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Imipramine hydrochloride, BioXtra, ≥99% (TLC)