Merck
  • The first fluorogenic assay for detecting a Baeyer-Villigerase activity in microbial cells.
The first fluorogenic assay for detecting a Baeyer-Villigerase activity in microbial cells.
Organic & biomolecular chemistry (2003-11-06)
María C Gutiérrez, Arthur Sleegers, Helen D Simpson, Véronique Alphand, Roland Furstoss
ABSTRACT
The first fluorogenic assay allowing for detection of microbial enzymes able to perform Baeyer-Villiger oxidation is described. This is based on the use of 4-oxopentyl umbelliferyl ether 1 as a fluorogenic substrate. When Baeyer-Villigerases active against this test ketone are present in the selected whole cells, 1 is transformed into 3-hydroxypropyl umbelliferyl ether 3, which, in a subsequent step, releases the fluorescent product umbelliferone. Different microorganisms, known to be endowed with Baeyer-Villigerase activity, were assayed.
Sigma-Aldrich
2-(3-Chloropropyl)-2-methyl-1,3-dioxolane, 97%