Merck
  • Identification and synthesis of new bicyclic acetals from the mountain pine beetle, Dendroctonus ponderosae Hopkins (Col.: Scol.).
Identification and synthesis of new bicyclic acetals from the mountain pine beetle, Dendroctonus ponderosae Hopkins (Col.: Scol.).
Bioorganic & medicinal chemistry (1996-03-01)
W Francke, F Schröder, P Philipp, H Meyer, V Sinnwell, G Gries
ABSTRACT
Head-space volatiles obtained from male mountain pine beetles, Dendroctonus ponderosae, were analyzed by coupled GC-MS and chiral gas chromatography. 5-Ethyl-7-methyl-6,8-dioxabicyclo[3.2.1]octane (6) was found as a new naturally occurring isomer of brevicomin (1). In addition, several stereoisomers of 7-ethyl-5-methyl-6,8-dioxabicyclo[3.2.1]octan-2-ol (11) and 1-(5-methyl-6,8-dioxabicyclo[3.2.1]octyl)ethanol (12) could be identified. Relative and absolute configurations of the compounds were determined by unambiguous syntheses, which are described.