Merck
  • Marginal fidelity and microleakage of porcelain veneers made by two techniques.
Marginal fidelity and microleakage of porcelain veneers made by two techniques.
The Journal of prosthetic dentistry (1992-01-01)
J A Sorensen, J M Strutz, S P Avera, D Materdomini
ABSTRACT
This study evaluated the marginal fidelity and microleakage of porcelain veneers made with the platinum foil and refractory die techniques. Maxillary incisors, matched for size and amount of enamel, were prepared with 0.5 mm uniform intraenamel reduction. The indirectly made veneers were etched, treated with silane, and luted with a composite resin, and the margins were finished and polished. The restored teeth were stored in 37 degrees C water, thermocycled 1000 times, stained with silver nitrate, embedded, sectioned buccolingually and mesiodistally, and measured at x250 magnification. The platinum foil veneers had significantly better vertical marginal fidelity but significantly more overcontouring than had the refractory die veneers. Universal microleakage at the tooth-composite resin interface and negligible microleakage at the porcelain-composite resin interface were observed. No relationship was found between the amount of vertical marginal opening and the amount of microleakage.
Product Description
Platinum, foil, light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Platinum, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 0.1m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Platinum, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, 0.1m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Platinum, foil, 15x15mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 50x50mm, thickness 0.1mm, as rolled, 99.99+%
Platinum, foil, 0.05m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Platinum, foil, 0.1m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 0.05m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Platinum, foil, 0.5m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Platinum, foil, 0.05m coil, thickness 0.030mm, coil width .7mm, as rolled, 99.95%
Platinum, foil, 15mm disks, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Platinum, foil, 30x30mm, thickness 0.2mm, as rolled, 99.95%
Platinum, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Platinum, foil, not light tested, 50x50mm, thickness 0.003mm, as rolled, 99.95%
Platinum, foil, 4mm disks, thickness 0.0075mm, as rolled, 99.95%
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Platinum, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95%
Platinum, foil, 8mm disks, thickness 0.01mm, as rolled, 99.95%
Platinum, foil, 6mm disks, thickness 0.005mm, as rolled, 99.95%
Platinum, foil, 0.2m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Platinum, foil, not light tested, 50x50mm, thickness 0.0015mm, as rolled, 99.95%
Platinum, foil, 8mm disks, thickness 0.004mm, as rolled, 99.95%
Platinum, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%
Platinum, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99.95%