Merck
Synthesis of amphidinolide E C10-C26 fragment.
Organic letters (2008-10-08)
Jorge Esteban, Anna M Costa, Jaume Vilarrasa
ABSTRACT
The key C10-C26 fragment in a total synthesis of (-)-amphidinolide E has been prepared from an oxolane-containing C10-C17 segment (9, derived from L-glutamic acid) via a Julia-Kocienski reaction with aldehyde 3, followed by a Sharpless AD to obtain the desired diol. The C22-C26 fragment was installed by means of an efficient Suzuki-Molander coupling, with an organotrifluoroborate reagent (4, arising from a cross-metathesis reaction between a vinylboronate and 2-methyl-1,4-pentadiene).