Merck
  • Clinical application and quantitative evaluation of generator-produced copper-62-PTSM as a brain perfusion tracer for PET.
Clinical application and quantitative evaluation of generator-produced copper-62-PTSM as a brain perfusion tracer for PET.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (1994-12-01)
H Okazawa, Y Yonekura, Y Fujibayashi, S Nishizawa, Y Magata, K Ishizu, F Tanaka, T Tsuchida, N Tamaki, J Konishi
ABSTRACT
Copper-62-pyruvaldehyde bis(N4-methylthiosemicarbazone) copper II (62Cu-PTSM) has been proposed as a generator-produced positron-emitting tracer for perfusion imaging. To evaluate the characteristics of 62Cu-PTSM as a cerebral perfusion tracer, brain PET images of 62Cu-PTSM were compared with cerebral blood flow (CBF). Following an intravenous injection of 62Cu-PTSM, a serial dynamic PET scan was performed for 10 min with arterial sampling in 10 subjects. CBF was measured by 15O-labeled water before the 62Cu-PTSM study. Dynamic PET scan with octanol-extracted arterial input function indicated the presence of significant back-diffusion of 62Cu-PTSM from the brain within 3 min after injection, followed by stable activity from 3 to 10 min. Comparison with 15O-water PET demonstrated less contrast between high- and low-flow regions in 62Cu-PTSM image and a nonlinear relationship of flow and 62Cu-PTSM uptake, which suggests the underestimation of CBF in high-flow regions due to the existence of back-diffusion. Although 62Cu-PTSM can be used widely for evaluation of brain perfusion with PET, kinetic analysis and correction may be needed to quantify regional CBF.
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%