Merck
  • Determination of lignans and isoflavonoids in human female plasma following dietary supplementation.
Determination of lignans and isoflavonoids in human female plasma following dietary supplementation.
The Journal of endocrinology (1994-08-01)
M S Morton, G Wilcox, M L Wahlqvist, K Griffiths
ABSTRACT
Plasma levels of the lignans enterodiol and enterolactone, and also the isoflavonic phyto-oestrogens daidzein, equol and genistein, are reported for postmenopausal Australian women consuming a traditional diet supplemented with linseed, soya flour or clover sprouts. Analysis was performed by gas chromatography-mass spectrometry, after enzymatic hydrolysis and ion-exchange chromatography. Following linseed supplementation, combined levels of enterolactone and enterodiol reached 500 ng/ml, whereas after soya flour or clover sprouts the respective concentrations of equol, daidzein and genistein reached 43, 312 and 148 ng/ml. Not all subjects were able to produce equol from daidzein. The possible relationship and role of these weak dietary oestrogens as restraining factors in the development of hormone-dependent cancers in Asian populations is discussed.
Product Description
Supelco
Daidzein, analytical standard
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Supelco
Genistein, analytical standard
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)