Merck
Radioimmunoassay for 1,25-dihydroxyvitamin D in serum or plasma.
Clinical chemistry (1985-01-01)
A P De Leenheer, R M Bauwens
ABSTRACT
For this sensitive RIA for 1 alpha,25-dihydroxyvitamin D, we used antibodies to 1 alpha,25-dihydroxycholecalciferol-3-hemisuccinate conjugated to bovine serum albumin, raised in rabbits. These antibodies show a high affinity for 1 alpha,25-dihydroxyvitamin D3 but cross react with other vitamin D metabolites as well. Extraction and liquid chromatography are required to isolate the 1 alpha,25-dihydroxyvitamin D from human serum or plasma with benzene (analytical recovery, 96%) and chromatographed by "high-performance" liquid chromatography on a silica column. The sensitivity of the assay is 2 pg/tube. The between-assay CV is 12.6% (means = 61.8 ng/L, n = 27) and the within-assay CV is 7.4% (means = 61.8 ng/L, n = 15). The reference interval, obtained from data on serum of healthy persons, is 51.8 +/- 15.1 ng/L (means +/- SD, n = 91), with no sex-related difference. Patients with chronic renal failure have lower concentrations in serum, and patients suffering from calcium nephrolithiasis have increased concentrations. Our assay offers the advantage of a relatively short prepurification procedure and the production of large amounts of stable gamma-globulins that can be used for several years without loss of binding properties.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland