Merck
The use of perfluorocarbon liquids in vitreous surgery.
Bulletin de la Societe belge d'ophtalmologie (1990-01-01)
C Claes, R Zivojnovic
ABSTRACT
Perfluoro-octane is used as an intraoperative tool for manipulation of the retina during vitreous surgery for retinal detachment in cases of proliferative vitreoretinopathy (PVR), giant tears and large peripheral retinotomies. One case of a macular hole combined with a peripheral one was also treated in this way. Perfluoro-octane seems, in our experience, a useful instrument for intraoperative atraumatic flattening of the retinal in those complicated cases with largely detached retinas.
Sigma-Aldrich
Octane, anhydrous, ≥99%
Supelco
Octane, analytical standard
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Octane, reagent grade, 98%