Merck
  • Associations between urinary phthalate monoesters and thyroid hormones in pregnant women.
Associations between urinary phthalate monoesters and thyroid hormones in pregnant women.
Human reproduction (Oxford, England) (2007-08-21)
Po-Chin Huang, Pao-Lin Kuo, Yue-Liang Guo, Pao-Chi Liao, Ching-Chang Lee
ABSTRACT
Maternal hypothyroidism during pregnancy can cause adverse effects in the fetus. Scientific evidence has shown that probable thyroid-like function of some phthalates in vitro and in vivo, and phthalates exposure, can begin in utero. This study investigated the association between phthalate exposure and thyroid hormones in pregnant women. Serum and spot urine samples were collected from 76 Taiwanese pregnant women at second trimester. Thyroid hormones, including thyroid-stimulating hormone (TSH), triiodothyronine (T(3)), thyroxine (T(4)) and free T(4) (FT(4)) were analysed in serum samples, and five urinary phthalate monoesters, including mono butyl phthalate (MBP), monoethyl phthalate (MEP) and mono ethylhexyl phthalate (MEHP), were measured. Urinary MBP, MEP and MEHP, the median levels of which were 81.8, 27.7 and 20.6 ng/ml, respectively, were the predominant substances in the urinary phthalate monoesters. Significant mild negative correlations were found between T(4) and urinary MBP (R = -0.248, P < 0.05), and between FT(4) and urinary MBP (R = -0.368, P < 0.05). After adjusting for age, BMI and gestation, urinary MBP levels showed negative associations with FT(4) and T(4) (FT(4): beta = -0.110, P < 0.001; T(4): beta=-0.112, P = 0.003). Exposure to di-n-butyl phthalate (DBP) may affect thyroid activity in pregnant women, but how DBP affects thyroid function is unclear. Further studies are needed to elucidate the mechanism of action and to investigate whether any other factors related to DBP exposure alter the thyroid function.
Product Description
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Estriol 16α-(β-D-glucuronide), ≥97%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-erythro-Dihydrosphingosine, ≥98%
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Estriol, meets USP testing specifications
Supelco
Progesterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®