Merck
  • Dose-dependent inhibition of platelet function by acetaminophen in healthy volunteers.
Dose-dependent inhibition of platelet function by acetaminophen in healthy volunteers.
Anesthesiology (2005-09-30)
Edward Munsterhjelm, Nina M Munsterhjelm, Tomi T Niemi, Olavi Ylikorkala, Pertti J Neuvonen, Per H Rosenberg
ABSTRACT
Acetaminophen (paracetamol) is widely used for postoperative analgesia. Its mechanism of action is inhibition of prostaglandin synthesis in the central nervous system, and acetaminophen is traditionally not considered to influence platelet function. The authors studied the dose-dependent inhibition of platelet function by acetaminophen in healthy volunteers. Thirteen healthy male volunteers (aged 19-26 yr) were given placebo or 15, 22.5, or 30 mg/kg acetaminophen intravenously in a double-blind, crossover study. Ten and 90 min after infusion, platelet function was assessed by photometric aggregometry and by measuring release of thromboxane B2, analgesia by cold pressor test, and plasma acetaminophen concentrations by high-performance liquid chromatography. When triggered with 500 microm arachidonic acid, median platelet aggregation (area under the curve) was 25.7, 22.8, 4.1, or 3.6 x 10 area units (P < 0.001) 10 min after placebo or 15, 22.5, or 30 mg/kg acetaminophen, respectively. An increasing concentration of arachidonic acid attenuated the antiaggregatory effect. After 90 min, platelet function was recovering. Release of thromboxane B2 was also dose-dependently inhibited by acetaminophen. Although plasma concentration of acetaminophen increased linearly with the dose, no analgesic effect was detected in the cold pressor test. Acetaminophen, which is a weak inhibitor of platelet cyclooxygenase 1, has a dose-dependent antiaggregatory effect. This property may become clinically significant in patients with intrinsic or drug-induced impairment of hemostasis.
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder