Merck
  • Differential acetylcholine and choline concentrations in the cerebrospinal fluid of patients with Alzheimer's disease and vascular dementia.
Differential acetylcholine and choline concentrations in the cerebrospinal fluid of patients with Alzheimer's disease and vascular dementia.
Chinese medical journal (2004-09-14)
Jian-ping Jia, Jian-min Jia, Wei-dong Zhou, Min Xu, Chang-biao Chu, Xin Yan, Yong-xin Sun
ABSTRACT
An important aspect of Alzheimer's disease (AD) is loss or impairment of cholinergic neurons. It is controversial whether there is a similar cholinergic impairment and cerebral deficit of acetylcholine (ACh) in the case of vascular dementia (VD). The purpose of this study was to explore the levels of ACh and choline (Ch) in the cerebrospinal fluid (CSF) of patients with AD and VD, and their possible relationship with cognitive impairment. Twenty-two AD patients, twenty-two VD patients, and twenty normal controls were recruited and scored with a Mini-Mental State Examination (MMSE). CSF concentrations of ACh and Ch were measured using high-performance liquid chromatography with an electrochemical detector (HPLC-ECD) and the results were then compared to cognitive status. ACh concentrations in CSF of AD patients [(10.7 +/- 5.1) nmol/L] and VD patients [(16.8 +/- 7.4) nmol/L] were both significantly lower than in controls [(34.5 +/- 9.0) nmol/L, t = 10.67, P < 0.001; t = 6.91, P < 0.001]. Both results correlated positively with MMSE scores (rs = 0.88 and rs = 0.85, respectively, P < 0.01). The CSF concentration of Ch was significantly higher in VD patients [(887.4 +/- 187.4) nmol/L] compared to AD patients [(627.6 +/- 145.1) nmol/L, t = 6.4, P < 0.001] and controls [(716.0 +/- 159.4) nmol/L, t = 4.2, P = 0.002]. CSF Ch concentration showed no difference between AD patients and normal controls, nor did it correlate with MMSE score in any of the three groups. The positive correlation between ACh deficit and cognitive impairment suggests that ACh is an important neurotransmitter for memory. The similar decrease in ACh concentration in AD and VD patients may imply a similar pathogenesis for the process of cognitive impairment involved in these two disorders. The elevated CSF levels of Ch in VD patients compared to AD patients may be useful diagnostically. Cholinesterase inhibitors may be helpful not only for AD patients, but also for VD patients.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
L-Proline, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
L-Proline, 99%, FCC, FG
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
L-Proline, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%