Merck
  • Markers of dopamine metabolism in Parkinson's disease. The Parkinson Study Group.
Markers of dopamine metabolism in Parkinson's disease. The Parkinson Study Group.
Neurology (1992-11-01)
P A LeWitt, M P Galloway, W Matson, P Milbury, M McDermott, D K Srivastava, D Oakes
ABSTRACT
We used two analytic methods (a multichannel coulometric electrode array with high-performance liquid chromatography, and gas chromatography-mass spectrophotometry) to measure CSF dopamine (DA) and its metabolites in mildly affected, unmedicated subjects with Parkinson's disease (PD). The mean (+/- SD) concentration of homovanillic acid (HVA), the most abundant product of DA turnover, was 164.57 +/- 95.05 nM. As sequential aliquots of CSF were collected from the first to 23rd ml, CSF HVA concentration almost doubled. After HVA, 3-O-methyldopa (3-O-MD) was the next most abundant compound. The summed concentrations of 3-O-MD, 3,4-dihydroxyphenylacetic acid, 3-methoxytyramine, DA, DA-3-sulfate, homovanillol, and levodopa (LD) amounted to 12.6% of HVA. Concentrations of the DA metabolites did not correlate to a variety of indices of PD severity. The presence of LD and 3-O-MD may be indicators of DA synthesis and possibly could reflect compensatory processes among surviving dopaminergic neurons of the PD brain.
Product Description
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
S-(5′-Adenosyl)-L-homocysteine, crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Cystine
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
Chromium, chips, 99.995% trace metals basis
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Nickel, powder, <50 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Strontium, granular, 99% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Titanium, wire, diam. 0.81 mm, 99.7% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Strontium, dendritic pieces, purified by distillation, 99.99% trace metals basis