Merck
  • Amino acids and biogenic amines in cerebrospinal fluid of patients with Parkinson's disease.
Amino acids and biogenic amines in cerebrospinal fluid of patients with Parkinson's disease.
Neurochemical research (2003-07-02)
S Engelborghs, B Marescau, P P De Deyn
ABSTRACT
To study changes in amino acid metabolism and biogenic amines in Parkinson's disease, we set up a prospective study and measured biogenic amines, their main metabolites, and 22 different amino acids, in cerebrospinal fluid of Parkinson's disease patients (n = 24) and age-matched controls (n = 30). A trend toward higher dopamine levels in Parkinson's disease patients was interpreted as an effect of treatment with levodopa and/or selegiline. Significantly lower concentrations of the dopamine metabolite 3,4-dihydroxyphenylacetic acid in the Parkinson's disease group might reflect dopaminergic cell loss. Our results revealed decreased serotonin catabolism that was interpreted as an effect of treatment with selegiline. Whereas all amino acid levels were unchanged, taurine was significantly lower in Parkinson's disease patients. Studies showed that taurine exerts a trophic action on the central nervous system. In this view, decreased taurine in a neurodegenerative disorder as Parkinson's disease deserves attention.
Product Description
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Supelco
L-Methionine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
SAFC
L-Methionine
SAFC
L-Isoleucine
SAFC
Taurine