Merck
  • Prenatal diagnosis of molybdenum cofactor deficiency and isolated sulfite oxidase deficiency.
Prenatal diagnosis of molybdenum cofactor deficiency and isolated sulfite oxidase deficiency.
Prenatal diagnosis (2003-01-21)
Jean L Johnson
ABSTRACT
Molybdenum cofactor deficiency and isolated sulfite oxidase deficiency are autosomal recessive inborn errors of metabolism with severe neurological symptoms resulting from a lack of sulfite oxidase activity. The deficiencies can be diagnosed prenatally by monitoring sulfite oxidase activity in chorionic villus sampling (CVS) tissue. In those families in which the specific defects have been identified, diagnosis can be achieved by mutation analysis or linkage studies directed at affected genes. These include MOCS1, MOCS2 or GEPH, in cases of molybdenum cofactor deficiency, or SUOX in patients with isolated sulfite oxidase deficiency.
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis