Merck
  • N(epsilon)-(gamma-L-glutamyl)-L-lysine (GGEL) is increased in cerebrospinal fluid of patients with Huntington's disease.
N(epsilon)-(gamma-L-glutamyl)-L-lysine (GGEL) is increased in cerebrospinal fluid of patients with Huntington's disease.
Journal of neurochemistry (2001-12-12)
T M Jeitner, M B Bogdanov, W R Matson, Y Daikhin, M Yudkoff, J E Folk, L Steinman, S E Browne, M F Beal, J P Blass, A J Cooper
ABSTRACT
Pathological-length polyglutamine (Q(n)) expansions, such as those that occur in the huntingtin protein (htt) in Huntington's disease (HD), are excellent substrates for tissue transglutaminase in vitro, and transglutaminase activity is increased in post-mortem HD brain. However, direct evidence for the participation of tissue transglutaminase (or other transglutaminases) in HD patients in vivo is scarce. We now report that levels of N(epsilon)-(gamma-L-glutamyl)-L-lysine (GGEL)--a 'marker' isodipeptide produced by the transglutaminase reaction--are elevated in the CSF of HD patients (708 +/- 41 pmol/mL, SEM, n = 36) vs. control CSF (228 +/- 36, n = 27); p < 0.0001. These data support the hypothesis that transglutaminase activity is increased in HD brain in vivo.
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
L-(+)-Lactic acid solution, 30% in H2O (by weight)
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Sigma-Aldrich
L-(+)-Lactic acid, BioXtra, ≥98% (titration)
Sigma-Aldrich
L-(+)-Lactic acid, ≥98%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-(+)-Lactic acid, 80%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%