- Simultaneous determination of trimipramine and desmethyl- and hydroxytrimipramine in plasma and red blood cells by capillary gas chromatography with nitrogen-selective detection.
Simultaneous determination of trimipramine and desmethyl- and hydroxytrimipramine in plasma and red blood cells by capillary gas chromatography with nitrogen-selective detection.
Journal of chromatography (1993-03-05)
N Jourdil, B Pinteur, F Vincent, C Marka, G Bessard
PMID8458903
ABSTRACT
A simple procedure is described that permits the simultaneous determination of trimipramine and its two major metabolites, desmethyl- and hydroxytrimipramine, in human plasma or red blood cells (RBCs) at therapeutic concentrations. The extracted biological fluids are injected into a capillary gas chromatograph with an OV-1 fused-silica column coupled to a nitrogen-phosphorus-selective detector. The limit of determination for trimipramine is 3 ng/ml and for that desmethyl- and hydroxytrimipramine is 4 ng/ml. The method permits the RBC/plasma ratios to be determined and to be correlated with the clinical response.
Product Description
Supelco
N-Desmethyltrimipramine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®