Merck
Stimulation of aflatoxin biosynthesis by lipophilic epoxides.
Journal of general microbiology (1983-06-01)
C Fanelli, A A Fabbri, E Finotti, S Passi
ABSTRACT
Epoxy fatty acids added to the culture media either with the inoculum or at the end of exponential growth phase stimulated aflatoxin production by toxigenic strains of Aspergillus flavus and Aspergillus parasiticus. This effect did not appear when the unsaturated fatty acids used for the synthesis of the epoxides and the polyhydroxyacids (which can be considered to be derived from the opening of the oxirane ring) replaced the epoxides in the culture media. No significant differences were detected in the lipid fractions (diglycerides, sterols, triglycerides, free fatty acids, sterol esters) extracted from the mycelia grown in the presence of any of the fatty acid derivates.