Merck
  • [Determination of acetylcholine by inhibition of the specific binding of tritiated cis-methyldioxolane: application to the study of the effects of acetylcholine accumulation in a murine neuroblastoma culture].
[Determination of acetylcholine by inhibition of the specific binding of tritiated cis-methyldioxolane: application to the study of the effects of acetylcholine accumulation in a murine neuroblastoma culture].
Comptes rendus des seances de la Societe de biologie et de ses filiales (1984-01-01)
G Blanchet, G Lallement, D Baubichon, S Mavet
ABSTRACT
The radioreceptor assay for acetylcholine (ACh) is based on the ability of the ACh to compete with the specific binding of [3H] cis-methyldioxolane to muscarinic receptors of rat cerebral cortex membranes. The technique described was used to measure ACh levels in NS 20 cells treated with an organophosphorus compound. The down regulation of muscarinic acetylcholine receptors of neuroblastoma cells observed is probably related to ACh accumulation.
Sigma-Aldrich
2-Methyl-1,3-dioxolane, 97%
Pricing and availability is not currently available.