- The preliminary evaluation of degradation of substance P(SP) fragment's analogue less than Glu SP6-11 in the subcellular fractions from different areas of rat brain.
The preliminary evaluation of degradation of substance P(SP) fragment's analogue less than Glu SP6-11 in the subcellular fractions from different areas of rat brain.
Peptidase(s) activity of different subcellular fractions isolated from cortex, hippocampus, midbrain, thalamus with hypothalamus, cerebellum and medulla oblongata exerted against less than Glu SP6-11 (3H-Phen8) was evaluated in "low-ionic" and similar (in composition) to both extracellular and intracellular conditions. The incubation of less than Glu SP6-11 with different fractions leaves the hexapeptide undegraded in the studied conditions in most cases. Peptidases activity results in the formation of the first of all C-terminal and exceptionally "internal" labelled products. Labelled N-terminal products were not seen. The most effective degradation in vitro of less than Glu SP6-11 takes place, in the majority of cases, in "low ionic" conditions when compared to those similar to extra or intracellular ones. The biggest total (per 1 g of wet mass) and specific activities against less than Glu SP6-11 can be shown in the hippocampus areas.