Merck
  • Proton magnetic resonance spectroscopy (H-MRS) in chronic schizophrenia. A single-voxel study in three regions involved in a pathogenetic theory.
Proton magnetic resonance spectroscopy (H-MRS) in chronic schizophrenia. A single-voxel study in three regions involved in a pathogenetic theory.
The neuroradiology journal (2013-07-19)
F Granata, G Pandolfo, S Vinci, C Alafaci, N Settineri, R Morabito, A Pitrone, M Longo
ABSTRACT
The cognitive dysmetria theory suggests a disconnectivity between the dorsolateral prefrontal cortex, thalami and vermis to explain the pathophysiology of schizophrenia. This study investigated the metabolic integrity of this neurologic circuit in patients with schizophrenia using proton magnetic resonance spectroscopy (H-MRS). Twenty-two patients with schizophrenia and twelve control subjects were studied. Metabolites concentrations were evaluated by a single-voxel technique in the prefrontal cortex, thalami and vermis. To our knowledge, this is the first H-MRS experience with concomitant evaluation of these regions in schizophrenic patients. We found no significant statistical difference in N-AA, Cho and Cr absolute concentrations and N-AA/Cho, N-AA/Cr and Cho/Cr ratios between the schizophrenic patients and control group. At the vermis, we found a constant spectrum with low levels of N-AA and higher levels of Cho and Cr. Our experience does not clearly support or refute the cognitive dysmetria theory. The consistency of metabolic findings in the cerebellar vermis could represent an important datum, highlighting the specificity of metabolic and functional activity in this region.
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Creatine, anhydrous