Merck
  • Reduced sulfation of chondroitin sulfate but not heparan sulfate in kidneys of diabetic db/db mice.
Reduced sulfation of chondroitin sulfate but not heparan sulfate in kidneys of diabetic db/db mice.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2013-06-13)
Trine M Reine, Frøy Grøndahl, Trond G Jenssen, Elin Hadler-Olsen, Kristian Prydz, Svein O Kolset
ABSTRACT
Heparan sulfate proteoglycans are hypothesized to contribute to the filtration barrier in kidney glomeruli and the glycocalyx of endothelial cells. To investigate potential changes in proteoglycans in diabetic kidney, we isolated glycosaminoglycans from kidney cortex from healthy db/+ and diabetic db/db mice. Disaccharide analysis of chondroitin sulfate revealed a significant decrease in the 4-O-sulfated disaccharides (D0a4) from 65% to 40%, whereas 6-O-sulfated disaccharides (D0a6) were reduced from 11% to 6%, with a corresponding increase in unsulfated disaccharides. In contrast, no structural differences were observed in heparan sulfate. Furthermore, no difference was found in the molar amount of glycosaminoglycans, or in the ratio of hyaluronan/heparan sulfate/chondroitin sulfate. Immunohistochemical staining for the heparan sulfate proteoglycan perlecan was similar in both types of material but reduced staining of 4-O-sulfated chondroitin and dermatan was observed in kidney sections from diabetic mice. In support of this, using qRT-PCR, a 53.5% decrease in the expression level of Chst-11 (chondroitin 4-O sulfotransferase) was demonstrated in diabetic kidney. These results suggest that changes in the sulfation of chondroitin need to be addressed in future studies on proteoglycans and kidney function in diabetes.
Product Description
Chondroitin sulfate sodium (marine), European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Chondroitin sulfate sodium, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Collagen Antibody, Type IV, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Chondroitin 4 Sulfate Antibody, clone BE-123, ascites fluid, clone BE-123, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Heparan Sulfate Proteoglycan (Perlecan) Antibody, clone A7L6, clone A7L6, from rat
Pricing and availability is not currently available.