Merck
  • Cobalt complexes as artificial hydrogenases for the reductive side of water splitting.
Cobalt complexes as artificial hydrogenases for the reductive side of water splitting.
Biochimica et biophysica acta (2013-05-22)
William T Eckenhoff, William R McNamara, Pingwu Du, Richard Eisenberg
ABSTRACT
The generation of H2 from protons and electrons by complexes of cobalt has an extensive history. During the past decade, interest in this subject has increased as a result of developments in hydrogen generation that are driven electrochemically or photochemically. This article reviews the subject of hydrogen generation using Co complexes as catalysts and discusses the mechanistic implications of the systems studied for making H2. This article is part of a Special Issue entitled: Metals in Bioenergetics and Biomimetics Systems.
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Cobalt, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.015mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%
Cobalt, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%
Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 8mm disks, thickness 0.025mm, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.0125mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.02mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Cobalt, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.004mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.002mm, temporary acrylic support, 99.9%
Cobalt, foil, 8mm disks, thickness 0.02mm, 99.9%