Merck
  • HOPE-preservation of paraffin-embedded sputum samples--a new way of bioprofiling in COPD.
HOPE-preservation of paraffin-embedded sputum samples--a new way of bioprofiling in COPD.
Respiratory medicine (2013-01-15)
Frauke Pedersen, Sebastian Marwitz, Sophie Seehase, Anne-Marie Kirsten, Peter Zabel, Ekkehard Vollmer, Klaus F Rabe, Helgo Magnussen, Henrik Watz, Torsten Goldmann
ABSTRACT
Induced sputum is a non-invasive sampling technique for the analysis of airway inflammation in various lung diseases and comprises valuable potential for the identification of biomarkers and therapeutic targets by molecular methods. In the context of biobanking with preservation of induced sputum samples for subsequent analyses we applied the HEPES-glutamic acid buffer-mediated organic solvent protection effect (HOPE)-technique for preparation of induced sputum samples. Induced sputum samples of 20 patients with moderate to severe chronic obstructive pulmonary disease (COPD) and 12 healthy controls were collected. Cell pellets of induced sputum samples were preserved with HOPE and subsequently embedded in paraffin. Immunostaining of paraffin-block sections for interleukin-8, interleukin-17, myeloperoxidase, matrixmetalloproteinase-9, CD68, and CD8 revealed distinct signals without antigen retrieval. Moreover, RNA was extracted and successfully used for transcription microarray analysis. Sputum samples preserved by the HOPE-technique display a tool to address scientific approaches in pulmonary research, which can enable the identification of new biomarkers and therapeutic targets in respiratory diseases.
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES sodium salt
SAFC
HEPES
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture