Merck
  • Single-step cloning of a hybridoma producing a monoclonal antibody against a target protein.
Single-step cloning of a hybridoma producing a monoclonal antibody against a target protein.
Analytical biochemistry (2012-11-14)
Hidefumi Orii, Takeshi Yamaguchi, Kenji Watanabe
ABSTRACT
We have developed a simple and time-saving method to identify hybridoma clones producing an antibody against a target protein among a large number of hybridomas in a single step. This method is very useful as the primary screening tool for hybridomas producing monoclonal antibodies if several micrograms of the target protein is available.
Sigma-Aldrich
Collodion solution, for microscopy, 2% in amyl acetate
Sigma-Aldrich
Collodion solution, 4-8% in ethanol/diethyl ether