Merck
  • Protein-induced structural evolution of silver sulfide at the nanoscale: from hollow particles to solid spheres.
Protein-induced structural evolution of silver sulfide at the nanoscale: from hollow particles to solid spheres.
Nanoscale (2012-06-28)
Jun Chen, Yifei Kong, Jiajia Ji, Jing Ruan, Kan Wang, Feng Gao, Daxiang Cui
ABSTRACT
Herein, a novel structural transformation of Ag(2)S nanoparticles from hollow particles to solid spheres is reported. The features of these structures are identified through a set of characterizations based on which the formation mechanism is also investigated.
Sigma-Aldrich
Silver sulfide, 99.9% trace metals basis