Merck
  • Synthesis, characterization and biological evaluation of some novel 17-isoxazoles in the estrone series.
Synthesis, characterization and biological evaluation of some novel 17-isoxazoles in the estrone series.
Steroids (2012-05-23)
Dóra Kovács, Zalán Kádár, Gergő Mótyán, Gyula Schneider, János Wölfling, István Zupkó, Eva Frank
ABSTRACT
Regioselective 1,3-dipolar cycloadditions of different aryl nitrile oxides to mestranol were carried out to furnish novel steroidal 17α-isoxazoles in good to excellent yields. Copper(I) was found to be an efficient catalyst, accelerating the intermolecular ring-closures and leading exclusively to 3,5-disubstituted isoxazoles. The yields of the cycloadducts, however, were influenced by the substituents on the aromatic moiety of the 1,3-dipoles. Moreover, dehydration of the primary products resulted in the corresponding Δ(16,17)exo-heterocyclic derivatives. All the synthesized compounds were subjected to in vitro pharmacological studies of their antiproliferative effects relative to three human malignant cell lines (HeLa, MCF7 and A2780).
Sigma-Aldrich
Mestranol, 99%