Merck
  • Polyuric-polydipsic syndrome in a pediatric case of non-glucocorticoid remediable familial hyperaldosteronism.
Polyuric-polydipsic syndrome in a pediatric case of non-glucocorticoid remediable familial hyperaldosteronism.
Endocrine journal (2012-03-27)
Alessandro Mussa, Roberta Camilla, Silvia Monticone, Francesco Porta, Daniele Tessaris, Francesca Verna, Paolo Mulatero, Silvia Einaudi
ABSTRACT
Familial hyperaldosteronism (FH) encompasses 3 types of autosomal dominant hyperaldosteronisms leading to inheritable hypertension. FH type II (FH-II), undistinguishable from sporadic hyperaldosteronism, represents the most frequent cause of inheritable hypertension and is believed to only manifest in adults. FH-III is a severe variety of PA resistant to pharmacotherapy and recently demonstrated to be caused by mutations in the gene encoding the potassium channel KCNJ5. In this report, we describe a FH pediatric patient, remarkable both for age at onset and unusual presentation: a two-years old girl with polyuric-polydipsic syndrome and severe hypertension, successfully treated with canrenone and amiloride. The girl had severe hypertension, hypokalemia, hypercalciuria, suppressed renin activity, high aldosterone, and unremarkable adrenal imaging. FH type I was ruled out by glucocorticoid suppression test, PCR test for CYP11B1/CYP11B2 gene, and urinary 18-oxo-cortisol and 18-hydroxy-cortisol excretion, which was in FH-II range. In spite of a clear-cut FH-II phenotype, the girl and her mother were found to harbor a FH-III genotype with KCNJ5 mutation (c.452G>A). Treatment with canrenone was started, resulting in prompt normalization of electrolytes and remission of polyuric-polydypsic syndrome. The addition of amiloride led to a complete normalization of blood pressure. This report expands the phenotypic spectrum of FH-III to a milder end, mimiking FH-II phenotype demonstrating that pharmacotherapy may be effective. This also implies that FH-II/III should be considered in the differential diagnosis of hypertensive children and, perhaps, that the offspring of patients with hyperaldosteronism should be screened for hypertension.
Product Description
Canrenone, European Pharmacopoeia (EP) Reference Standard