Merck
  • Methadone determination in puparia and its effect on the development of Lucilia sericata (Diptera, Calliphoridae).
Methadone determination in puparia and its effect on the development of Lucilia sericata (Diptera, Calliphoridae).
Forensic science international (2011-02-16)
Matthias Gosselin, Vincent Di Fazio, Sarah M R Wille, Maria del Mar Ramírez Fernandez, Nele Samyn, Benoit Bourel, Pierre Rasmont
ABSTRACT
This paper describes a sensitive UPLC-MS/MS method for quantification of methadone and 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP) in single empty puparial case of Lucilia sericata. Larvae were reared on substrates spiked with different concentrations of methadone (0-4 μg/g). Methadone was quantified in puparia reared on high concentrated substrates (0.8-4 μg/g). The major metabolite of methadone (EDDP) was not detected, confirming rapid elimination of metabolites by the larvae before pupation. The effects of methadone on the development of L. sericata were also investigated. No effect on sex ratio was detected. A significant difference was calculated for emerged adults but no trends could be observed. Concerning the developmental curve, a significant difference was observed between control and high methadone concentrations using the Kolmogorov-Smirnov test.
Product Description
Supelco
EDDP perchlorate solution, 1.0 mg/mL in methanol (as pyrolinium), ampule of 1 mL, certified reference material, Cerilliant®